Mueller-Hinton Agar
Supplier: IPM SCIENTIFIC, INC.
880017
470180-638EA 35.6 USD
470180-638
Mueller-Hinton Agar
Media for Microbiology Dehydrated Media for Microbiology
For testing antimicrobial susceptibility of microorganisms by agar diffusion method.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 tubes.