Bile Esculin Agar
Supplier: IPM SCIENTIFIC, INC.
880010
470180-632EA 30.85 USD
470180-632
Bile Esculin Agar
Media for Microbiology Dehydrated Media for Microbiology
For differential isolation and presumptive identification of group D streptococcus.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 tubes.