Dehydrated Media for Luria Broth
Supplier: IPM SCIENTIFIC, INC.
880009
470180-630EA 30.9 USD
470180-630
Dehydrated Media for Luria Broth
Media for Microbiology Dehydrated Media for Microbiology
Premeasured. Complete nutrient source for growing E. coli cultures.
Sterile, ready-to-use media. Note: Perishable. Please specify delivery date.