Ward's® Live Grassfrogs (Rana pipiens)
Supplier: CLAUDE LAMBERT
878200 878223
470180-586PK 97.95 USD
470180-586 470180-588 470180-590 470180-598 470180-602 470180-604
Ward's® Live Grassfrogs (Rana pipiens)
Specimens Live Specimens Animals - Living Material
Grass frogs are low maintenance and seasonally available, typically from September to July.
  • Great for In-Class Observation
  • Low-Maintenance
  • Also Known as Leopard Frogs
Keep in an aquarium or terrarium with both wet and dry areas and feed crickets and/or earthworms.
Ordering information: This kit includes living or perishable materials. They may not be available in all states. Check with your local department of agriculture.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
CANADIAN CUSTOMERS: Please note that due to Canadian import restrictions, frogs can no longer be packed with sphagnum moss.