Ward's® Drosophila Culturing Set
Supplier: Avantor
470180-400EA 19.95 USD
470180-400
Ward's® Drosophila Culturing Set
Specimens Live Specimens Animals - Living Material
Readily Observed Strain Requires No Magnification
Use this unique culturing set as a convenient introduction to genetics; students can perform a simple monohybrid cross to yield a 3:1 winged: wingless ratio. Females have red eyes with yellow bodies, males have white eyes with gray bodies, making them easy to distinguish. Cultures supplied are yellow-forked-attached and white, and yellow-forked-attached and white apterous. The set comes with WARD’S Drosophila Culture Guide and cultures for up to 15 setups.