Ward's® Live Sarcophaga Bullata Fly Larvae and Pupae
Supplier: Avantor
876355
470177-084EA 7.95 USD
470177-084 470180-392 470180-394 470180-472
Ward's® Live Sarcophaga Bullata Fly Larvae and Pupae
Specimens Live Specimens Animals - Living Material
Sarcophaga bullata are gray flesh flies. These flies are reared on decaying flesh and pupate quickly, within 5 to 7 days from birth, to illustrate complete metamorphosis in Diptera.
The (uninfected) pupae can serve as the host organism for emerging Nasonia larvae. The pupae can be stored under refrigeration for several weeks, effectively halting their life cycle and assuring a fresh supply of food for the Nasonia larvae.
Ordering information: Please specify a delivery date during checkout.
To request a specific instar, please contact us ten days in advance.
Delivery information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Packaging: A large, vigorous culture is shipped in a shatterproof plastic vial.