Aeolosoma
Supplier: CONNECTICUT VALLEY BIO SUPPLY
L29
470180-286EA 9.95 USD
470180-286 470180-288
Aeolosoma
Specimens Live Specimens Aquaria - Living Material
Aeolosoma sp. A small, freshwater Oligochaete, Aeolosoma is characterized by colorful oil droplets in its body. It’s easy to culture large quantities of this microscopic organism for use in regeneration experiments: Simply add hay and wheat grains to hay medium. Culture instructions are included. Size: 2–3 mm.