Ward's® Live Pond Snails
Supplier: WATERSCAPES AQUATIC NURSERY
874140 0 874142
470177-316PK 11.95 USD
470177-316 470177-318 470180-270
Ward's® Live Pond Snails
Specimens Live Specimens Aquaria - Living Material
Create a balanced ecosystem in your aquarium with the addition of these small, freshwater algae eaters. They reproduce rapidly, laying their eggs on the glass sides of the aquarium.
  • Key member of a balanced ecosystem
  • Remove algae and debris from your aquarium
  • Great for hands-on activities
Ordering information: These products contain living or perishable materials.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.