Ward's® Preying Protist Study Set
Supplier: Avantor
470180-204KT 17.95 USD
470180-204
Ward's® Preying Protist Study Set
Specimens Live Specimens Protists - Living Material
Observe Two Protists Hunting Included Prey
  • Study preying protists
  • Includes two types
  • Manual included
Simply add the Paramecium provided and watch the Chaos and Didinium specimens stalk, capture, and assimilate them. The set of three includes Chaos, Didinium, and Paramecium (prey).
**Please see the product resources tab for "Working with Protozoa" manual.