Ward's® Live Algae Classification Set
Supplier: Avantor
470179-938KT 70.95 USD
470179-938
Ward's® Live Algae Classification Set
Specimens Live Specimens Protists - Living Material
Examples of eight different types of algae, grouped by the color of their plastids.
Eight algae types are included in this set, each with a different plastid type, giving them a different color.
Bangia (Rhodophyte), Chlorella (Chlorophyte), Ectocarpus (Phaeophyte), Euglena (Euglenophyte), Peridinium (Dinoflagellate), Nitzschia (Bacillaria), Synura (Chrysophyte), Vaucheria (Xanthophyte).