Navicula
Supplier: Ward's Science
NAVI30/35
470179-846EA 9.95 USD
470179-846
Navicula
Specimens Live Specimens Protists - Living Material
Freshwater. Unicellular; pennate, with bilateral symmetry. Basic culture solution.
Characterized by shells (tests) made of silica, in two parts.