Ward's® Oil-Degrading Microbes Set
Supplier: Avantor
470179-704KT 52.95 USD
470179-704
Ward's® Oil-Degrading Microbes Set
Specimens Live Specimens Bacteria - Living Material
Study the Microbes Used in the Bioremediation of Oil Spills.
  • Useful in the process of cleaning up oil spills
This unique set features naturally occurring microbes that have an affinity for degrading petroleum products.
Penicillium sp. and Pseudomonas sp. are supplied in a convenient freeze-dried form, along with along with the media needed to rehydrate and incubate them.
The set also comes with the Working with Bacteria manual.
Ordering information: These products contain living or perishable materials.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.