Saprolegnia ferax, Tube Culture
Supplier: Ward's Science
156270
470179-666EA 13 USD
470179-666
Saprolegnia ferax, Tube Culture
Specimens Live Specimens Protists - Living Material
Water mold and fish parasite. Demonstrates sporangia and oospores. Grows on wheat seed, cucumber seed, or hemp seed in water environment. Reproduces sexually and asexually. Incubation temperature 25°C.
Cell walls made of cellulose; feeds by extending hyphae into host's tissue.