Ward's® Fungi-Like Protist Set
Supplier: Avantor
470179-630KT 49.95 USD
470179-630
Ward's® Fungi-Like Protist Set
Specimens Live Specimens Protists - Living Material
Organisms with Characteristics of Two Kingdoms
Eukaryotic and mobile like protists, yet with reproductive structures like those of fungi, these unique protists make interesting subjects to study. The set of three comes with a Dictyostelium discoideum (cellular slime mold) demonstration plate, Physarum polycephalum (plasmodial slime mold) demonstration plate, and Saprolegnia ferax (water mold) tube culture, as well as the Working with Protozoa manual.