Ward's® Antibiotic Resistance Kit
Supplier: Avantor
470179-628KT 111.95 USD
470179-628
Ward's® Antibiotic Resistance Kit
Educational Materials Biology Educational Materials Genetics Learning Activities
Using penicillin-impregnated discs on two plates, students grow one lawn each of two common, naturally occurring pathogen bacteria.
  • Students Compare Bacteria with and without Natural Resistance
  • Two Common Bacteria Included
  • Material for Five Lab Groups Included
  • Instructions Included
  • Penicillin-Impregnated Discs on Each Plate
The bacteria with natural resistance covers the entire plate with growth.
The bacteria without resistance shows a zone of inhibition (a ring of no growth around the disc).
Canada: Containment Level 2 – PHAC License required.
Ordering information: It includes Escherichia coli and Staphylococcus epidermidis tube cultures, material for five lab groups, and instructions.
Delivery information: This kit includes perishable materials. Please specify delivery date.
Caution: Note: Living cultures are provided to reduce the laboratory preparation time. They can be stored in the refrigerator for up to 3 months before use.