Ward's® Bacteria Forms Mixture
Supplier: Avantor
470179-596KT 23.5 USD
470179-596
Ward's® Bacteria Forms Mixture
Specimens Live Specimens Bacteria - Living Material
Three Morphology Types in One Tube
Students can now compare all three morphology types without creating separate slides. Tube cultures: Bacillus subtilis (rod), Kocuria rhizophila (cocci), and Aquaspirillum serpens (spirillum). Supplied in tryptic soy broth. The set of three includes an Explano-Mount, which is a microscope slide with an illustrated guide that highlights all the important information on the slide, and the Working with Bacteria manual.