Phycomyces blakesleeanus (+)
Supplier: Ward's Science
156180
470179-214EA 13.95 USD
470179-214
Phycomyces blakesleeanus (+)
Specimens Live Specimens Fungi - Living Material
Zygomycete. Mating strain, cross with Phycomyces blakesleeanus (-) to produce zygospores. Incubation temperature 25°C. Potato dextrose agar growth medium.