Chromobacterium violaceum
Supplier: Ward's Science
154931A
470179-192EA 13.95 USD
470179-192
Chromobacterium violaceum
Specimens Live Specimens Bacteria - Living Material
Found in fresh water. Chromogenesis; blue to purple pigments produced. Motile. Rods. Gram negative reaction. Aerobic. Incubation temperature 30°C. Do not refrigerate because it cannot survive in temperatures below 25°C. Nutrient broth growth medium. Note: Pathogen.
Packaged in a 16 x 125 mm tubes. Call our Customer Service for availability of demo plates and other special orders.
  • Biosafety Level 2
  • Canada: Containment Level 2 - PHAC License required