Ward's® Live Streptomyces griseus Culture
Supplier: Avantor
470179-182EA 13.95 USD
470179-182
Ward's® Live Streptomyces griseus Culture
Specimens Live Specimens Bacteria - Living Material
Produces streptomycin. Branching mycelium and coenocytic hyphae. 
  • Biosafety Level: 1
  • Motility: Negative
  • Morphology: Filamentous rods
  • Gram Reaction: Positive
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.