Ward's® Live Streptococcus gallolyticus Culture
Supplier: Avantor
470179-168EA 13.95 USD
470179-168
Ward's® Live Streptococcus gallolyticus Culture
Specimens Live Specimens Bacteria - Living Material
Isolated from the alimentary tract of cows. Lancefield group D, gamma hemolysis. May be pathogenic in humans. 
  • Biosafety Level: 1
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Negative
  • Morphology: Cocci small chains
  • Gram Reaction: Positive
  • Respiration: Aerobic
  • Hemolysis: Gamma
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: This organism may be pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.