Ward's® Live Clostridium beijerinckii Culture
Supplier: Avantor
470179-110EA 13.95 USD
470179-110
Ward's® Live Clostridium beijerinckii Culture
Specimens Live Specimens Bacteria - Living Material
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
  • Biosafety Level: 1
  • Gram Stain: Positive
  • Cell Morphology: Rods with spores
  • Motility: Positive
  • Respiration: Anaerobic
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living orMore Product Information