Ward's® Live Serratia liquefaciens Culture
Supplier: AMERICAN TYPE CULTURE COLL.
35551
470179-098EA 13.95 USD
470179-098
Ward's® Live Serratia liquefaciens Culture
Specimens Live Specimens Bacteria - Living Material
Colorless, most widely dispersed of the Serratia species. 
  • Biosafety Level: 1
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Positive
  • Morphology: Rods
  • Gram Reaction: Negative
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.