Ward's® Live Bacillus licheniformis Culture
Supplier: AMERICAN TYPE CULTURE COLL.
14580
470179-060EA 13.95 USD
470179-060
Ward's® Live Bacillus licheniformis Culture
Specimens Live Specimens Bacteria - Living Material
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
  • Biosafety Level: 1
  • Gram Stain: Positive
  • Cell Morphology: Rods with spores
  • Respiration: Aerobic
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.