Ward's® Live Coliphage Set
Supplier: Avantor
470179-040KT 50.95 USD
470179-040
Ward's® Live Coliphage Set
Specimens Live Specimens Viruses - Living Material
Practice Techniques Employed in Genetics and Biotechnology Studies.
The phages in this set are recommended for introducing students to handling techniques of virulent microbial viruses.
Set Contains: T4 (wild) and T4r (rapid lysis) with tube culture of host E. coli B and a teacher’s manual.
Ordering information: These products contain living or perishable materials.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.