Ward's® Animal Tissue Staining Lab Activity
Supplier: Avantor
470178-398KT
122.49
USD
470178-398
Ward's® Animal Tissue Staining Lab Activity
Chemicals for Science Education
Complete Fixing and Staining Package
- For Cell and Tissue Culture Preparations
- Ample supplies for numerous staining attempts
- Time Required: ~60 minutes
With enough materials to fix, stain, and mount numerous cell tissue culture preparations grown on glass slides, this kit is all you’ll need for students to learn fixation and staining techniques by actually performing the procedures. It comes with enough materials to perform up to 100 tissue culture slide preparations.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)