Pulpvinyl Bakelite Jar Caps
Supplier: Qorpak
CAP-00216 5015/12 CAP-00281 CAP-00293
470178-006CS 396.3 USD
470178-006 470178-008 470178-010 470178-012 470178-016
Pulpvinyl Bakelite Jar Caps
Closures Bottle Closures
Pulpvinyl liners resist most chemical reagents. Packages of 12.