Cork Slide
Supplier: Avantor
470177-680EA 6.7 USD
470177-680
Cork Slide
Slides Prepared Slides
Buoyant plant material
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
General structure of the outer tissue of Spanish oak. The walls around air spaces reminded Robert Hooke of small rooms in a monastery called “cells”.