Iron-Deficiency Anemia Slide
Supplier: Avantor
470177-660EA 6.65 USD
470177-660
Iron-Deficiency Anemia Slide
Slides Prepared Slides
Illustrates red blood cell characteristics of the deficiency
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Peripheral blood. Characterized by small, pale red blood cells.