Adult Scalp, Unpigmented Slide
Supplier: Avantor
470177-642EA 11.1 USD
470177-642
Adult Scalp, Unpigmented Slide
Slides Prepared Slides
Examine the epidermis of the head
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Shows epidermis, hair follicles, sebaceous glands, and other structures.