Cerebellum, Mammal Slide
Supplier: Avantor
470177-606EA 7.95 USD
470177-606
Cerebellum, Mammal Slide
Slides Prepared Slides
Illustrates Structures of the Brain
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Demonstrates molecular and granular layers of the cortex and Purkinje cells.