Epithelium, Composite Slide
Supplier: Avantor
470177-560EA 8.75 USD
470177-560
Epithelium, Composite Slide
Slides Prepared Slides
Compare and contrast epithelium types
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Sections of stratified squamous, columnar, and cuboidal epithelium on the same slide.