Mold Types, Composite Slide
Supplier: Avantor
470177-438EA 11.95 USD
470177-438
Mold Types, Composite Slide
Specimens Live Specimens Fungi - Living Material
Observe the differences between common mold types
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Aspergillus, Penicillium, and Rhizopus on the same slide, clearly contrasting sporangia and conidia.