Ward's® Live Termite (Reticulitermes sp.)
Supplier: DENNIS CATO
876460
470177-330PK 35.95 USD
470177-330
Ward's® Live Termite (Reticulitermes sp.)
Specimens Live Specimens Animals - Living Material
This species of termite is small and delicate bodied. It will follow a pen trail which mimics pheromones.
  • Demonstrate pheromone attraction in your classroom
  • Extremely easy to maintain in the laboratory or classroom
  • No reproductive termites are sent with the culture (we do not send queens)
This subterranean termite is smaller than Zootermopsis (also available), and also has distinct castes with workers, soldiers. Winged reproductive forms and occasionally secondary reproductive forms develop in the event of the queen’s death.
Ordering information: These products contain living or perishable materials.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Ward's holds valid USDA permits allowing us to ship to most continental US states. Please call Customer Service for permit details.