Ward's® Gram Stain Bacterial Suspension Kit
Supplier: Avantor
470177-256KT 20.99 USD
470177-256
Ward's® Gram Stain Bacterial Suspension Kit
Educational Materials Biology Educational Materials Microbiology Learning Activities
Culture Common Gram-Positive and Gram-Negative Bacteria
Use this handy kit in conjunction with our Gram Stain Kit for a complete exercise that covers culturing to Gram-staining identification. With this kit, students culture both a Gram-positive rod and a Gram-negative rod in preparation for staining. It comes with a freeze-dried mixed culture of Bacillus, culture materials, and instructions. Note: Includes perishable materials. Please specify delivery date.
Canada: Containment Level 2 – PHAC License required.