Thistle Tube Polypropylene Funnel, Bel-Art
Supplier: Bel-Art
F14751-0000_SE
470177-142EA 10.8 USD
470177-142
Thistle Tube Polypropylene Funnel, Bel-Art
Funnels Filtration Funnels Filter Funnels
This unbreakable plastic thistle tube is ideal for the high school laboratory. It has a two piece construction with the stem tightly fitting into the funnel. Stem O.D.: 6mm. Length: 290mm. Package of 12.