Colon Slide
Supplier: Avantor
470177-128EA 6.49 USD
470177-128
Colon Slide
Slides Prepared Slides
Illustrates a Segment of the Digestive Tract
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Through the large intestine, showing absence of villi and changes in muscular layers. Goblet cells are numerous.