Introductory Bacteria Slides
Supplier: Avantor
470177-040KT 79.99 USD
470177-040
Introductory Bacteria Slides
Slides Prepared Slides
Compare and contrast bacterial morphology
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Slides are supplied in a durable storage box.
Ordering information: Set includes Azotobacter chroococcum (sm); Spirillum volutans (sm); Agrobacterium tumefaciens (sm); Bacillus anthracis (sm); Escherichia coli (sm); Mycobacterium tuberculosis (sm); Salmonella enteritidis (sm); Staphylococcus aureus (sm); Treponema sp., (sm); Root nodules (cs).