Osteogenesis, Endochondral Ossification Slide
Supplier: Avantor
470176-956EA 9.9 USD
470176-956
Osteogenesis, Endochondral Ossification Slide
Slides Prepared Slides
Fetal skeletal development
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal long bone. Extensive deposition of bony spicules with both per- and endochondral osteoblasts and osteoclasts.