Taenia pisiformis, Mature Proglottid Slide
Supplier: Avantor
470176-918EA 8.25 USD
470176-918
Taenia pisiformis, Mature Proglottid Slide
Slides Prepared Slides
Endoparasitic Tapeworm
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mature proglottid of Taenia pisiformis, showing uterus, ovary, testes, sperm duct, and genital pore.