Cross Colored Threads Slide
Supplier: Avantor
470176-898EA 6.35 USD
470176-898
Cross Colored Threads Slide
Slides Prepared Slides
Whole mount
  • Teaches basic microscopy skills and lens characteristics
  • Rigorous quality control standards
For teaching students to focus the microscope. Yellow on top, followed by blue, with red on the bottom. Threads intersect at one point.