Syringa, Leaf Slide
Supplier: Avantor
470176-892EA 6.95 USD
470176-892
Syringa, Leaf Slide
Slides Prepared Slides
Illustrates characteristics of a temperate, dicot leaf
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Lilac. A typical mesophytic dicot leaf.