Preserved Onion Root Tips
Supplier: Avantor
470176-794EA 32.95 USD
470176-794
Preserved Onion Root Tips
Specimens Preserved Specimens Preserved Plant Materials
Excellent for slide making, or just viewing the various stages of mitosis.
  • Onion tips are preserved in alcohol
  • Use only what you need, and save the rest for later
Your students will gain valuable micro technique and staining experience by creating their own microscope slides of these onion root tips that have been specially grown, cut and fixed at maximum mitotic activity.