Live Paramecium bursaria
Supplier: CONNECTICUT VALLEY BIO SUPPLY
L2C
470176-722EA 9.95 USD
470176-722 470176-724
Live Paramecium bursaria
Specimens Live Specimens Protists - Living Material
Paramecium bursaria is a ciliate that contains mutualistic algae called Zoochlorella. The algae lives in the cytoplasma of the Paramecium and provides it with food. Paramecium bursaria grows in Dilute Alfalfa Medium (Hay Medium) or Soil Water Medium. Requires light for growth.