Ward's® Live Gloeocapsa Culture
Supplier: Ward's Science
GLOE30/35
470176-682EA 9.95 USD
470176-682 470176-684
Ward's® Live Gloeocapsa Culture
Specimens Live Specimens Bacteria - Living Material
This colonial cyanobacteria is structured as several cells bound together in a gelatinous "mucilaginous sheath".
Commonly found in masses on wet rocks in aquatic or semi-aquatic habitats.
Genus: Gloeocapsa Species: Not specified