Cosmarium
Supplier: Ward's Science
152140
470176-678EA 9.95 USD
470176-678
Cosmarium
Specimens Live Specimens Protists - Living Material
Freshwater. Unicellular; desmid. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.