Ward's® Live Chlorella pyrenoidosa Culture
Supplier: Ward's Science
152069
470176-674EA 9.95 USD
470176-674 470176-676
Ward's® Live Chlorella pyrenoidosa Culture
Specimens Live Specimens Protists - Living Material
This species of freshwater algae is commonly used for research on photosynthesis.
  • Excellent for studying photosynthesis
  • Rich in cellulose
It is characterized by green plastids and cell walls composed of cellulose. This species has a pyrenoid, which is the organelle that acts as the center of carbon dioxide fixation.
Genus: Chlorella Species: pyrenoidosa