Live Amoeba proteus cultures
Supplier: Avantor
870390 0
470176-446PK 9.95 USD
470176-446 470176-570 470176-572 470180-124 470189-294
Live Amoeba proteus cultures
Specimens Live Specimens Protists - Living Material
Demonstrate amoeba structure, locomotion, and feeding behavior.
  • Demonstrate amoeba structure, locomotion, and feeding behavior
  • ReadySlides are prepared for you you and offer a convenient, ready-to-go option
  • Vital Stained cultures allow students to better visualize organism structures
Amoeba proteus is a great sarcodine for demonstrating amoebae structure. They use pseudopods for both locamotion and feeding. Amoeba feed on Chilomonas, or other small protozoa, which they phagocytize with their pseudopodia. Amoeba cultures are supplied in Amoeba Media with a rice grain and food (Chilomonas).
Ordering information: These products contain living or perishable materials. Please specify a delivery date.