Microbiology Morphology Slides
Supplier: Avantor
470176-204KT 68.5 USD
470176-204
Microbiology Morphology Slides
Slides Prepared Slides
Investigate the differences between microorganisms
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
This set is designed to show the basic differential shapes between bacteria and also between bacteria and fungi. An excellent set to demonstrate the morphology of micro-organisms supplied in a durable box.
Ordering information: Set includes Penicillum (wm); Yeast, budding (wm); Capsules of bacteria sm); Typical coccus (sm); Typical spirillum (sm); Bacteria types (sm); Bacillus in chains (sm); Typical mixed bacteria (sm); Spirochaetes (sm); Treponema pallidium (sm); Typical bacillus (sm); Staphylococcus epidermidis (sm).