Clay Triangles
Supplier: United Scientific Supplies
CPT020
470176-154EA 3.2 USD
470176-154
Clay Triangles
Tripods
Clay pipe triangle for holding items over a flame.
  • Heat resistant clay pipes
Galvanized, 16-gauge iron wire with plain clay-pipe stems.