Test Tube Brush
Supplier: PRECISION MOULDED POLYMERS LIMITED
1350765060000
470175-994EA 4 USD
470175-994
Test Tube Brush
Brushes
Ideal for cleaning test tubes and other tubing apparatus.
Nylon filament mounted in heavy guage twisted wire and completed with double radial tufted end. Synthetic material and nylon bristles provide effective cleaning over the entire bottom of the test tube.
Packaging: Packaged within a heat-sealed polybag.